missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic Acid Monohydrate 98.0+%, TCI America™

Catalog No. p-7118311
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B003025G 25 g
B0030500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B003025G Supplier TCI America Supplier No. B003025G
Only null left

Chemical Identifiers

CAS 26158-00-9
Molecular Formula C6H8O4S
Molecular Weight (g/mol) 176.186
MDL Number MFCD00149976
InChI Key MVIOINXPSFUJEN-UHFFFAOYSA-N
PubChem CID 15619088
IUPAC Name benzenesulfonic acid;hydrate
SMILES C1=CC=C(C=C1)S(=O)(=O)O.O

Specifications

Quantity 25 g
UN Number 2585
Formula Weight 158.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Benzenesulfonic Acid Monohydrate
EINECSNumber (3)-1928
RTECSNumber DB4200000
TSCA Yes