accessibility menu, dialog, popup

UserName

SOOHOH2O

CAS RN 26158-00-9

IUPAC Name: benzenesulfonic acid hydrate
Synonyms: benzenesulfonic acid hydrate
Molecular Weight (g/mol): 176.19
Molecular Formula: C6H8O4S
InChi Key: MVIOINXPSFUJEN-UHFFFAOYSA-N
SMILES: O.OS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results