Learn More
Benzenesulfonic acid, 90%, technical
CAS: 98-11-3 | C6H6O3S | 158.17 g/mol
Supplier: Thermo Scientific Chemicals 159021000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Benzenesulfonic acid | |
| 88.0 | |
| 45.0°C to 50.0°C | |
| >100°C | |
| 90% | |
| C6H6O3S | |
| MFCD00011689 | |
| 11,26 | |
| benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech | |
| SRSXLGNVWSONIS-UHFFFAOYSA-N | |
| benzenesulfonic acid | |
| 7371 | |
| 158.17 | |
| Technical |
| 98-11-3,7664-93-9 | |
| 100.0 | |
| Yellow to Brown | |
| Authentic | |
| Glass bottle | |
| C6H5SO3H | |
| 100 g | |
| 15,1071 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in benzene,insoluble in ether and carbon disulfide | |
| C1=CC=C(C=C1)S(=O)(=O)O | |
| 158.17 | |
| CHEBI:64455 | |
| 90% | |
| Damp Crystalline Solid or Fused Mass |
Chemical Identifiers
| 98-11-3 | |
| 158.17 | |
| SRSXLGNVWSONIS-UHFFFAOYSA-N | |
| 7371 | |
| benzenesulfonic acid |
| C6H6O3S | |
| MFCD00011689 | |
| benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech | |
| CHEBI:64455 | |
| C1=CC=C(C=C1)S(=O)(=O)O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 202-638-7
RUO – Research Use Only