missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic acid, 90%, technical

Catalog No. p-3777365
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159020250 25 g Glass bottle
AC159021000 100 g Glass bottle
AC159020010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC159020250 Supplier Thermo Scientific Chemicals Supplier No. 159020250
Only null left

CAS: 98-11-3 | C6H6O3S | 158.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-11-3 , 7664-93-9
Molecular Formula C6H6O3S
Molecular Weight (g/mol) 158.17
MDL Number MFCD00011689
InChI Key SRSXLGNVWSONIS-UHFFFAOYSA-N
Synonym benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
PubChem CID 7371
ChEBI CHEBI:64455
IUPAC Name benzenesulfonic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 45.0°C to 50.0°C
Color Yellow to Brown
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula C6H5SO3H
Quantity 25 g
Beilstein 11,26
Merck Index 15,1071
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in benzene,insoluble in ether and carbon disulfide
Formula Weight 158.17
Percent Purity 90%
Grade Technical
Physical Form Damp Crystalline Solid or Fused Mass
Chemical Name or Material Benzenesulfonic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 202-638-7

RUO – Research Use Only