missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonanilide 98.0+%, TCI America™

Catalog No. p-7118310
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B002925G 25 g
B0029500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B002925G Supplier TCI America Supplier No. B002925G
Only null left

Chemical Identifiers

CAS 1678-25-7
Molecular Formula C12H11NO2S
Molecular Weight (g/mol) 233.29
MDL Number MFCD00035919
InChI Key XAUGWFWQVYXATQ-UHFFFAOYSA-N
Synonym benzenesulfonanilide, benzoylsulfanilide, benzenesulfoanilide, benzensulfonanilide, benzenesulfanilide, benzenesulfonamide, n-phenyl, n-phenylbenzenesulphonamide, phenyl phenylsulfonyl amine, n-phenyl-benzenesulfonamide, chembl83148
PubChem CID 74296
IUPAC Name N-phenylbenzenesulfonamide
SMILES O=S(=O)(NC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 110°C
Color White
Quantity 25 g
Formula Weight 233.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfonanilide
EINECSNumber (3)-3781
RTECSNumber DB3500000
TSCA No