missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonamide, 98+%

Catalog No. p-62896
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148490050 5 g Glass bottle
AC148491000 100 g Plastic bottle
AC148495000 500 g Plastic bottle
AC148490025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC148490050 Supplier Thermo Scientific Chemicals Supplier No. 148490050
Only null left

CAS: 98-10-2 | C6H7NO2S | 157.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-10-2
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00007930
InChI Key KHBQMWCZKVMBLN-UHFFFAOYSA-N
Synonym benzenesulphonamide, benzosulfonamide, benzolsulfonamide, phenyl sulfonamide, benzene sulfonamide, phenylsulfonamide, m and b 7973, chembl27601, benzolsulfonamid, benzensulfonamide
PubChem CID 7370
IUPAC Name benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 151°C to 153°C
Color White
Flash Point 250°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SO2NH2
Quantity 5 g
Beilstein 11, 39
Solubility Information Solubility in water: 4.3g/L (16°C)
Formula Weight 157.19
Percent Purity 98+%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Benzenesulfonamide, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 202-637-1
RTECSNumber DA9380000
TSCA TSCA

RUO – Research Use Only