missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific™
$134.05 - $451.02
Chemical Identifiers
| CAS | 873-55-2 |
|---|---|
| Molecular Formula | C6H5NaO2S |
| Molecular Weight (g/mol) | 164.15 |
| MDL Number | MFCD00013135 |
| InChI Key | CHLCPTJLUJHDBO-UHFFFAOYSA-M |
| Synonym | sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate |
| PubChem CID | 2723840 |
| IUPAC Name | sodium;benzenesulfinate |
| SMILES | C1=CC=C(C=C1)S(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC154051000
|
N/A | 100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC154055000
|
N/A | 500g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H5NaO2S | |
| MFCD00013135 | |
| sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate | |
| sodium;benzenesulfinate |
Specifications
| 873-55-2 | |
| 100.0 | |
| 97% | |
| C6H5NaO2S | |
| MFCD00013135 | |
| 06,526 | |
| CHLCPTJLUJHDBO-UHFFFAOYSA-M | |
| sodium;benzenesulfinate | |
| 2723840 | |
| 97% |
| 96.0 | |
| Authentic | |
| Plastic bottle | |
| C6H5SO2Na | |
| 11, 2 | |
| sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate | |
| C1=CC=C(C=C1)S(=O)[O-].[Na+] | |
| 164.15 | |
| 164.15 | |
| Benzenesulfinic acid, sodium salt, 97% |
Safety and Handling
EINECSNumber : 212-842-8
TSCA : TSCA
RUO – Research Use Only