missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzenepentacarboxylic Acid 98.0+%, TCI America™
$1228.41 - $4911.79
Chemical Identifiers
| CAS | 1585-40-6 |
|---|---|
| Molecular Formula | C11H6O10 |
| Molecular Weight (g/mol) | 298.159 |
| MDL Number | MFCD00020281 |
| InChI Key | QNSOHXTZPUMONC-UHFFFAOYSA-N |
| PubChem CID | 15314 |
| IUPAC Name | benzene-1,2,3,4,5-pentacarboxylic acid |
| SMILES | C1=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Chemical Identifiers
| 1585-40-6 | |
| 298.159 | |
| QNSOHXTZPUMONC-UHFFFAOYSA-N | |
| benzene-1,2,3,4,5-pentacarboxylic acid |
| C11H6O10 | |
| MFCD00020281 | |
| 15314 | |
| C1=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Specifications
| 1585-40-6 | |
| C11H6O10 | |
| 5 g | |
| C1=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O | |
| 298.159 | |
| 298.16 | |
| Crystalline Powder |
| White | |
| MFCD00020281 | |
| QNSOHXTZPUMONC-UHFFFAOYSA-N | |
| benzene-1,2,3,4,5-pentacarboxylic acid | |
| 15314 | |
| ≥98.0% (HPLC,T) | |
| Benzenepentacarboxylic Acid |
Safety and Handling
TSCA : No