accessibility menu, dialog, popup

UserName

HOOOHOOHOHOOOHO

CAS RN 1585-40-6

IUPAC Name: benzene-1,2,3,4,5-pentacarboxylic acid
Synonyms: benzenepentacarboxylic acid
Molecular Weight (g/mol): 298.16
Molecular Formula: C11H6O10
InChi Key: QNSOHXTZPUMONC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(C(O)=O)=C(C(O)=O)C(C(O)=O)=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results