missing translation for 'onlineSavingsMsg'
Learn More

Benzenepentacarboxylic Acid 98.0+%, TCI America™

Catalog No. B09525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09525G 5 g
B095225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09525G Supplier TCI America Supplier No. B09525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1585-40-6
Molecular Formula C11H6O10
Molecular Weight (g/mol) 298.159
MDL Number MFCD00020281
InChI Key QNSOHXTZPUMONC-UHFFFAOYSA-N
PubChem CID 15314
IUPAC Name benzene-1,2,3,4,5-pentacarboxylic acid
SMILES C1=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 298.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzenepentacarboxylic Acid