missing translation for 'onlineSavingsMsg'
Learn More

Benzene-1,4-dithiocarboxamide, 97%

Catalog No. AAH5185703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5185703 1 g
AAH5185706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5185703 Supplier Thermo Scientific Chemicals Supplier No. H5185703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13363-51-4 | C8H8N2S2 | 196.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13363-51-4
Molecular Formula C8H8N2S2
Molecular Weight (g/mol) 196.286
MDL Number MFCD00046866
InChI Key USHPIZCRGQUHGN-UHFFFAOYSA-N
Synonym benzene-1,4-dithiocarboxamide, 1,4-benzenedicarbothioamide, benzene-1,4-bis carbothioamide
PubChem CID 2795175
IUPAC Name benzene-1,4-dicarbothioamide
SMILES C1=CC(=CC=C1C(=S)N)C(=S)N

Specifications

Quantity 1 g
UN Number UN2811
Formula Weight 196.28
Percent Purity 97%
Chemical Name or Material Benzene-1,4-dithiocarboxamide
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only