missing translation for 'onlineSavingsMsg'
Learn More

Benzbromarone 98.0+%, TCI America™

$104.38 - $346.71

Chemical Identifiers

CAS 3562-84-3
Molecular Formula C17H12Br2O3
Molecular Weight (g/mol) 424.088
MDL Number MFCD00078962
InChI Key WHQCHUCQKNIQEC-UHFFFAOYSA-N
Synonym benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
PubChem CID 2333
ChEBI CHEBI:3023
IUPAC Name (3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
SMILES CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B40991G
View Documents
TCI America
B40991G
1 g N/A
Only null left
 
B40995G
View Documents
TCI America
B40995G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3562-84-3
424.088
WHQCHUCQKNIQEC-UHFFFAOYSA-N
2333
(3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
C17H12Br2O3
MFCD00078962
benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
CHEBI:3023
CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br

Specifications

3562-84-3
White-Yellow
MFCD00078962
2811
WHQCHUCQKNIQEC-UHFFFAOYSA-N
(3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
2333
424.09
Crystalline Powder
152°C
C17H12Br2O3
1 g
benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br
424.088
CHEBI:3023
≥98.0% (GC,T)
Benzbromarone
Videos
Safety and Handling

Safety and Handling

RTECSNumber : OB1804200

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'