missing translation for 'onlineSavingsMsg'
Learn More

Benzbromarone, 98%

Catalog No. AC460900050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460900050 5 g
AC460900010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460900050 Supplier Thermo Scientific Chemicals Supplier No. 460900050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3562-84-3 | C17H12Br2O3 | 424.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3562-84-3
Molecular Formula C17H12Br2O3
Molecular Weight (g/mol) 424.08
InChI Key WHQCHUCQKNIQEC-UHFFFAOYSA-N
Synonym benzbromarone, benzbromaron, desuric, urinorm, normurat, uricovac, minuric, exurate, hipurik, azubromaron
PubChem CID 2333
ChEBI CHEBI:3023
IUPAC Name (3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
SMILES CCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)Br)O)Br

Specifications

Melting Point 152.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in hot methanol
Formula Weight 424.08
Percent Purity 98%
Chemical Name or Material Benzbromarone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 222-630-7

RUO – Research Use Only