missing translation for 'onlineSavingsMsg'
Learn More

Benzanilide 98.0+%, TCI America™

Catalog No. p-7118302
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B001625G 25 g
B0016500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B001625G Supplier TCI America Supplier No. B001625G
Only null left

Chemical Identifiers

CAS 93-98-1
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.237
MDL Number MFCD00003069
InChI Key ZVSKZLHKADLHSD-UHFFFAOYSA-N
Synonym benzanilide, n-benzoylaniline, benzamide, n-phenyl, n-phenyl-benzamide, benzoic acid anilide, unii-ak1b12366o, phenyl-n-benzamide, benzanilid, benzoylanilide, n-benzoyl aniline
PubChem CID 7168
IUPAC Name N-phenylbenzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2

Specifications

Melting Point 166°C
Color White
Quantity 25 g
Formula Weight 197.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzanilide