accessibility menu, dialog, popup

UserName

CAS RN 93-98-1

IUPAC Name: N-phenylbenzamide
Synonyms: benzanilide
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: ZVSKZLHKADLHSD-UHFFFAOYSA-N
SMILES: O=C(NC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)