missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzalmalononitrile 98.0+%, TCI America™
$152.86 - $305.70
Chemical Identifiers
| CAS | 2700-22-3 |
|---|---|
| Molecular Formula | C10H6N2 |
| Molecular Weight (g/mol) | 154.172 |
| MDL Number | MFCD00001855 |
| InChI Key | WAVNYPVYNSIHNC-UHFFFAOYSA-N |
| Synonym | benzylidenemalononitrile, 2-benzylidenemalononitrile, benzalmalononitrile, benzylidenemalonodinitrile, malononitrile, benzylidene, benzal-malonitril, benzylidene malononitrile, 2,2-dicyano-1-phenylethylene, 2-phenyl-1,1-dicyanoethylene, beta,beta-dicyanostyrene |
| PubChem CID | 17608 |
| IUPAC Name | 2-benzylidenepropanedinitrile |
| SMILES | C1=CC=C(C=C1)C=C(C#N)C#N |
Chemical Identifiers
| 2700-22-3 | |
| 154.172 | |
| WAVNYPVYNSIHNC-UHFFFAOYSA-N | |
| 17608 | |
| C1=CC=C(C=C1)C=C(C#N)C#N |
| C10H6N2 | |
| MFCD00001855 | |
| benzylidenemalononitrile, 2-benzylidenemalononitrile, benzalmalononitrile, benzylidenemalonodinitrile, malononitrile, benzylidene, benzal-malonitril, benzylidene malononitrile, 2,2-dicyano-1-phenylethylene, 2-phenyl-1,1-dicyanoethylene, beta,beta-dicyanostyrene | |
| 2-benzylidenepropanedinitrile |
Specifications
| 2700-22-3 | |
| White-Yellow | |
| MFCD00001855 | |
| 3439 | |
| WAVNYPVYNSIHNC-UHFFFAOYSA-N | |
| 2-benzylidenepropanedinitrile | |
| 17608 | |
| ≥98.0% (GC) | |
| Benzalmalononitrile |
| 85°C | |
| C10H6N2 | |
| 10 g | |
| benzylidenemalononitrile, 2-benzylidenemalononitrile, benzalmalononitrile, benzylidenemalonodinitrile, malononitrile, benzylidene, benzal-malonitril, benzylidene malononitrile, 2,2-dicyano-1-phenylethylene, 2-phenyl-1,1-dicyanoethylene, beta,beta-dicyanostyrene | |
| C1=CC=C(C=C1)C=C(C#N)C#N | |
| 154.172 | |
| 154.17 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : OO3500000
TSCA : No