missing translation for 'onlineSavingsMsg'
Learn More

Benzalmalononitrile 98.0+%, TCI America™

Catalog No. B094710G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B094710G 10 g
B094725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B094710G Supplier TCI America Supplier No. B094710G

Chemical Identifiers

CAS 2700-22-3
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.172
MDL Number MFCD00001855
InChI Key WAVNYPVYNSIHNC-UHFFFAOYSA-N
Synonym benzylidenemalononitrile, 2-benzylidenemalononitrile, benzalmalononitrile, benzylidenemalonodinitrile, malononitrile, benzylidene, benzal-malonitril, benzylidene malononitrile, 2,2-dicyano-1-phenylethylene, 2-phenyl-1,1-dicyanoethylene, beta,beta-dicyanostyrene
PubChem CID 17608
IUPAC Name 2-benzylidenepropanedinitrile
SMILES C1=CC=C(C=C1)C=C(C#N)C#N

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 10 g
UN Number 3439
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzalmalononitrile
RTECSNumber OO3500000
TSCA No