missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride, 95-104%, Spectrum™ Chemical

Catalog No. p-7157072
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
100 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Glass Bottle
Catalog No. Quantity Packaging
18600874 100 g Amber Glass Bottle
18600872 1 kg Glass Bottle
18600873 25 kg Fiber Drum
3 options

Catalog No. 18600874

Supplier: Spectrum Chemical Mfg Cor B3775100GM

Only null left

B3775, 8001-54-5

Spectrum™ Chemical Benzalkonium Chloride is used as a biocide, a phase transfer agent, and a cationic surfactant in the chemical industry. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 8001-54-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
SMILES [Cl-].C[N+](C)(*)CC1=CC=CC=C1

Specifications

CAS Max % 100%
Quantity 100 g
UN Number UN2923
Percent Purity 95 to 104%
Grade Reagent
Packaging Amber Glass Bottle