missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzaldehyde Phenylhydrazone 98.0+%, TCI America™
Supplier: TCI America B001425G
Specifications
| Benzaldehyde Phenylhydrazone | |
| 156°C | |
| MFCD00051318 | |
| JGOAZQAXRONCCI-SDNWHVSQSA-N | |
| N-[(E)-benzylideneamino]aniline | |
| 6436752 | |
| ≥98.0% (HPLC) |
| 588-64-7 | |
| C13H12N2 | |
| 25 g | |
| C1=CC=C(C=C1)C=NNC2=CC=CC=C2 | |
| 196.253 | |
| 196.25 | |
| Crystalline Powder |
Chemical Identifiers
| 588-64-7 | |
| 196.253 | |
| JGOAZQAXRONCCI-SDNWHVSQSA-N | |
| N-[(E)-benzylideneamino]aniline |
| C13H12N2 | |
| MFCD00051318 | |
| 6436752 | |
| C1=CC=C(C=C1)C=NNC2=CC=CC=C2 |
Safety and Handling
TSCA : Yes