missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzaldehyde p-Toluenesulfonylhydrazone 98.0+%, TCI America™
$162.17 - $466.02
Chemical Identifiers
| CAS | 1666-17-7 |
|---|---|
| Molecular Formula | C14H14N2O2S |
| Molecular Weight (g/mol) | 274.338 |
| MDL Number | MFCD00009644 |
| InChI Key | FZFLTDNAHASQQC-PTNGSMBKSA-N |
| Synonym | Benzaldehyde Tosylhydrazone |
| PubChem CID | 5397192 |
| IUPAC Name | N-[(Z)-benzylideneamino]-4-methylbenzenesulfonamide |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)NN=CC2=CC=CC=C2 |
Chemical Identifiers
| 1666-17-7 | |
| 274.338 | |
| FZFLTDNAHASQQC-PTNGSMBKSA-N | |
| 5397192 | |
| CC1=CC=C(C=C1)S(=O)(=O)NN=CC2=CC=CC=C2 |
| C14H14N2O2S | |
| MFCD00009644 | |
| Benzaldehyde Tosylhydrazone | |
| N-[(Z)-benzylideneamino]-4-methylbenzenesulfonamide |
Specifications
| 1666-17-7 | |
| White-Yellow | |
| MFCD00009644 | |
| Benzaldehyde Tosylhydrazone | |
| CC1=CC=C(C=C1)S(=O)(=O)NN=CC2=CC=CC=C2 | |
| 274.338 | |
| 274.34 | |
| Crystalline Powder |
| 130°C | |
| C14H14N2O2S | |
| 5 g | |
| FZFLTDNAHASQQC-PTNGSMBKSA-N | |
| N-[(Z)-benzylideneamino]-4-methylbenzenesulfonamide | |
| 5397192 | |
| ≥98.0% (HPLC,T) | |
| Benzaldehyde p-Toluenesulfonylhydrazone |
Safety and Handling
TSCA : No