missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzaldehyde Dimethyl Acetal 98.0+%, TCI America™
Supplier: TCI America B1197100ML
Specifications
| Benzaldehyde Dimethyl Acetal | |
| Colorless | |
| C9H12O2 | |
| 100 mL | |
| HEVMDQBCAHEHDY-UHFFFAOYSA-N | |
| dimethoxymethylbenzene | |
| 62375 | |
| ≥98.0% (GC) |
| 1125-88-8 | |
| 196°C | |
| MFCD00008491 | |
| benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca | |
| COC(C1=CC=CC=C1)OC | |
| 152.193 | |
| 152.19 | |
| Liquid |
Chemical Identifiers
| 1125-88-8 | |
| 152.193 | |
| HEVMDQBCAHEHDY-UHFFFAOYSA-N | |
| 62375 | |
| COC(C1=CC=CC=C1)OC |
| C9H12O2 | |
| MFCD00008491 | |
| benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca | |
| dimethoxymethylbenzene |
Safety and Handling
EINECSNumber : (3)-1152
RTECSNumber : CU5774000
TSCA : Yes