Learn More
Benzaldehyde dimethyl acetal, 99%
CAS: 1125-88-8 | C9H12O2 | 152.193 g/mol
Supplier: Thermo Scientific Chemicals A110090E
Description
Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.
Solubility
Insoluble in water. Soluble in alcohol.
Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents and acids.
Specifications
| Benzaldehyde dimethyl acetal | |
| 1.014 | |
| 69°C (156°F) | |
| C9H12O2 | |
| MFCD00008491 | |
| 2044501 | |
| Insoluble in water. Soluble in alcohol. | |
| COC(C1=CC=CC=C1)OC | |
| 152.193 | |
| 152.2 |
| 1125-88-8 | |
| 196°C | |
| Characteristic | |
| 1.4935 | |
| 2500 g | |
| benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca | |
| HEVMDQBCAHEHDY-UHFFFAOYSA-N | |
| dimethoxymethylbenzene | |
| 62375 | |
| 99% |
Chemical Identifiers
| 1125-88-8 | |
| 152.193 | |
| HEVMDQBCAHEHDY-UHFFFAOYSA-N | |
| 62375 | |
| COC(C1=CC=CC=C1)OC |
| C9H12O2 | |
| MFCD00008491 | |
| benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca | |
| dimethoxymethylbenzene |
Safety and Handling
GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
H227-H302
EINECSNumber : 214-413-0
RTECSNumber : CU5774000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only