missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde dimethyl acetal, 99%

Catalog No. AAA1100922
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1100922 100 g
AAA1100936 500 g
AAA110090E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1100922 Supplier Thermo Scientific Chemicals Supplier No. A1100922

CAS: 1125-88-8 | C9H12O2 | 152.193 g/mol

Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents and acids.

Chemical Identifiers

CAS 1125-88-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00008491
InChI Key HEVMDQBCAHEHDY-UHFFFAOYSA-N
Synonym benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
PubChem CID 62375
IUPAC Name dimethoxymethylbenzene
SMILES COC(C1=CC=CC=C1)OC

Specifications

Density 1.014
Boiling Point 196°C
Flash Point 69°C (156°F)
Odor Characteristic
Refractive Index 1.4935
Quantity 100 g
Beilstein 2044501
Solubility Information Insoluble in water. Soluble in alcohol.
Formula Weight 152.2
Percent Purity 99%
Chemical Name or Material Benzaldehyde dimethyl acetal
Show More Show Less
Hazard Category H227-H302
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 214-413-0
RTECSNumber CU5774000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only