missing translation for 'onlineSavingsMsg'
Learn More

Benzalazine 98.0+%, TCI America™

Catalog No. b000625g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
B000625G 25 g
1 options

Catalog No. B000625G

Supplier: TCI America B000625G

Only null left

Chemical Identifiers

CAS 588-68-1
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.26
MDL Number MFCD00016876
InChI Key CWLGEPSKQDNHIO-JOBJLJCHSA-N
Synonym 1,2-dibenzylidenehydrazine, n-z-benzylideneamino-1-phenylmethanimine, 1,2-di e-benzylidene hydrazine, e-n-e-benzylideneamino-1-phenylmethanimine
PubChem CID 5371793
IUPAC Name (E,E)-bis(phenylmethylidene)hydrazine
SMILES C(=N/N=C/C1=CC=CC=C1)\C1=CC=CC=C1

Specifications

Melting Point 93°C
Color Yellow
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzalazine
RTECSNumber CU7571500
TSCA Yes
Recommended Storage Refrigerator