missing translation for 'onlineSavingsMsg'
Learn More

Benzal Diacetate 95.0+%, TCI America™

Catalog No. p-7118298
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B001025G 25 g
B0010500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B001025G Supplier TCI America Supplier No. B001025G
Only null left

Chemical Identifiers

CAS 581-55-5
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00059206
InChI Key XRYSDRCNTMEYFH-UHFFFAOYSA-N
Synonym benzal diacetate, benzylidene diacetate, phenylmethylene diacetate, benzaldiacetate, methanediol, phenyl-, diacetate, benzaldiacetat, m-diacetoxymethylbenzene, diacetic acid benzylidene, phenylmethanediyl diacetate, phenylmethanediol diacetate
PubChem CID 68489
IUPAC Name [acetyloxy(phenyl)methyl] acetate
SMILES CC(=O)OC(C1=CC=CC=C1)OC(=O)C

Specifications

Melting Point 45°C
Color Yellow
Boiling Point 154°C
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material Benzal Diacetate