CAS RN 581-55-5
IUPAC Nor:
(acetyloxy)(phenyl)methyl acetate
Synonymes:
benzal diacetate
Poids moléculaire (g/mol):
208.21
Formule moléculaire:
C11H12O4
InChi Key:
XRYSDRCNTMEYFH-UHFFFAOYSA-N
SMILES:
CC(=O)OC(OC(C)=O)C1=CC=CC=C1
1
–
1
de
1
résultats
1
Benzal Diacetate 95.0+%, TCI America™
CAS: 581-55-5 Formule moléculaire: C11H12O4 Poids moléculaire (g/mol): 208.213 Numéro MDL: MFCD00059206 Clé InChI: XRYSDRCNTMEYFH-UHFFFAOYSA-N Synonyme: benzal diacetate,benzylidene diacetate,phenylmethylene diacetate,benzaldiacetate,methanediol, phenyl-, diacetate,benzaldiacetat,m-diacetoxymethylbenzene,diacetic acid benzylidene,phenylmethanediyl diacetate,phenylmethanediol diacetate CID PubChem: 68489 Nom IUPAC: [acetyloxy(phenyl)methyl] acetate SMILES: CC(=O)OC(C1=CC=CC=C1)OC(=O)C