missing translation for 'onlineSavingsMsg'
Learn More

Bentonite

CAS: 1302-78-9

Supplier:  Thermo Scientific Chemicals 447440025

 View more versions of this product

Catalog No. AC447440025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent, Lewis acid
Specifications

Specifications

Bentonite
Plastic bottle
15,1057
1302-78-9
2.5 kg
Solubility in water: insoluble. Other solubilities: insoluble in acids and common organic solvents

Chemical Identifiers

1302-78-9
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-108-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only