missing translation for 'onlineSavingsMsg'
Learn More

Bentonite, Powder, NF, Spectrum™ Chemical

Catalog No. 18600870
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
22.5 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600870 12 kg Fiber Drum
18600868 500 g Poly Bottle
18600869 2.5 kg Poly Pail
18600871 22.5 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600870 Supplier Spectrum Chemical Mfg Cor Supplier No. BE12512KG
Only null left

BE125, 1302-78-9

Spectrum™ Chemical Bentonite, Powder, NF is used as suspending and emulsifying agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1302-78-9
Molecular Formula Al2H2O12Si4
Molecular Weight (g/mol) 360.31
InChI Key KMDMICAGCNGGEB-UHFFFAOYSA-N
IUPAC Name tetrasilicon(4+) dialuminium(3+) hydrate undecaoxidandiide
SMILES O.[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Si+4].[Si+4].[Si+4].[Si+4]

Specifications

CAS Max % 100%
Loss on Drying 5.0-8%
Packaging Fiber Drum
Quantity 12 kg
Grade NF