missing translation for 'onlineSavingsMsg'
Learn More

Benserazide hydrochloride

Catalog No. AAJ6458103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6458103 1 g
AAJ6458106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6458103 Supplier Thermo Scientific Chemicals Supplier No. J6458103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of L-aromatic amino acid decarboxylase | CAS: 14919-77-8 | C10H16ClN3O5 | 293.704 g/mol

Inhibitor of L-aromatic amino acid decarboxylaseBenserazide hydrochloride is used as an antiparkinsonian. It is a peripheral decarboxylase inhibitor. It is used as an inhibitor for aromatic L-aminoacid decarboxylase. Further, it serves as an inhibitor of tyrosine hydroxylase (TH) and Tryptophan hydroxylase (TPH) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibitor of L-aromatic amino acid decarboxylaseBenserazide hydrochloride is used as an antiparkinsonian. It is a peripheral decarboxylase inhibitor. It is used as an inhibitor for aromatic L-aminoacid decarboxylase. Further, it serves as an inhibitor of tyrosine hydroxylase (TH) and Tryptophan hydroxylase (TPH) inhibitor.

Solubility
Soluble in water, dimethyl sulfoxide and methanol. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 14919-77-8
Molecular Formula C10H16ClN3O5
Molecular Weight (g/mol) 293.704
MDL Number MFCD00078571
InChI Key ULFCBIUXQQYDEI-UHFFFAOYSA-N
Synonym benserazide hydrochloride, benserazide hcl, 2-amino-3-hydroxy-n'-2,3,4-trihydroxybenzyl propanehydrazide hydrochloride, ccris 5092, 2'-2,3,4-trihydroxybenzyl-dl-serinohydrazide monohydrochloride, benzerazide hydrochloride, dl-serine 2, dl-serine 2-2,3,4-trihydroxybenzyl hydrazine hydrochloride, 2-amino-3-hydroxy-n'-2,3,4-trihydroxyphenyl methyl propanehydrazide hydrochloride
PubChem CID 26964
ChEBI CHEBI:31262
IUPAC Name 2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride
SMILES C1=CC(=C(C(=C1CNNC(=O)C(CO)N)O)O)O.Cl

Specifications

Melting Point 146°C to 148°C
Color White
Quantity 1 g
Sensitivity Light sensitive
Merck Index 14,1050
Solubility Information Soluble in water,dimethyl sulfoxide and methanol. Insoluble in ethanol and acetone.
Formula Weight 293.7
Physical Form Powder
Chemical Name or Material Benserazide hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-991-9
RTECSNumber VT9632300
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only