accessibility menu, dialog, popup

UserName

CAS RN 14919-77-8

IUPAC Name: hydrogen 2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide chloride
Synonyms: benserazide
Molecular Weight (g/mol): 293.7
Molecular Formula: C10H16ClN3O5
InChi Key: ULFCBIUXQQYDEI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC(CO)C(=O)NNCC1=CC=C(O)C(O)=C1O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)