missing translation for 'onlineSavingsMsg'
Learn More

Benserazide Hydrochloride 97.0+%, TCI America™

Catalog No. B41085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41085G 5 g
B41081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41085G Supplier TCI America Supplier No. B41085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14919-77-8
Molecular Formula C10H16ClN3O5
Molecular Weight (g/mol) 293.704
MDL Number MFCD00078571
InChI Key ULFCBIUXQQYDEI-UHFFFAOYSA-N
Synonym benserazide hydrochloride, benserazide hcl, 2-amino-3-hydroxy-n'-2,3,4-trihydroxybenzyl propanehydrazide hydrochloride, ccris 5092, 2'-2,3,4-trihydroxybenzyl-dl-serinohydrazide monohydrochloride, benzerazide hydrochloride, dl-serine 2, dl-serine 2-2,3,4-trihydroxybenzyl hydrazine hydrochloride, 2-amino-3-hydroxy-n'-2,3,4-trihydroxyphenyl methyl propanehydrazide hydrochloride
PubChem CID 26964
ChEBI CHEBI:31262
IUPAC Name 2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride
SMILES C1=CC(=C(C(=C1CNNC(=O)C(CO)N)O)O)O.Cl

Specifications

Melting Point 180°C
Quantity 5 g
Formula Weight 293.70
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benserazide Hydrochloride
RTECSNumber VT9632300
TSCA No