missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benedict's Solution, Quantitative, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Benedict's Solution |
| Name Note | Quantitative |
| CAS | 7732-18-5, 6132-04-3, 333-20-0, 497-19-8 |
| Assay Percent Range | 2.00 ± 0.01mg glucose/mL |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| pH | Alkaline |
| Quantity | 100 mL |