missing translation for 'onlineSavingsMsg'
Learn More

Bendroflumethiazide, MilliporeSigma™ Supelco™

Catalog No. 111015378
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015378 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015378 Supplier Merck Emd Millipore Supplier No. B57751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bendroflumethiazide belongs to the class of thiazide diuretics, widely used in enhancing diuresis. It is also used as an effective antihypertensive agent and in the treatment of edema caused by cardiac/renal failure and myocardial infarction. Its mode of action involves increasing the urine flow and the net renal excretion of both sodium and water. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 73-48-3
Physical Form Neat
Linear Formula C15H14F3N3O4S2
MDL Number MFCD00078963
Synonym 3-Benzyl-6-trifluoromethyl-7-sulfamoyl-3,4-dihydro-2 H-1,2,4-benzothiadiazine 1,1-dioxide
Type Analytical Standard
Quantity 1 g
Formula Weight 421.41
Grade Analytical Standard