missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bendazac, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 464920010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Bendazac | |
| C16H13N2NaO3 | |
| Solubility aq. soln.: insoluble. Other solubilities: soluble in chloroform, ethanol, acetone | |
| [Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12 | |
| 304.28 |
| 20187-55-7 | |
| 1 g | |
| GHFGHIKJGVMEFT-UHFFFAOYSA-M | |
| sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate | |
| 282.3 |
Chemical Identifiers
| 20187-55-7 | |
| 304.28 | |
| sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate |
| C16H13N2NaO3 | |
| GHFGHIKJGVMEFT-UHFFFAOYSA-M | |
| [Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12 |
Safety and Handling
GHS H Statement Harmful if swallowed.
GHS Signal Word: Warning
EINECSNumber : 243-569-2
RUO â Research Use Only