missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bendazac 97.0+%, TCI America™
$123.21 - $363.85
Chemical Identifiers
| CAS | 20187-55-7 |
|---|---|
| Molecular Formula | C16H14N2O3 |
| Molecular Weight (g/mol) | 282.299 |
| MDL Number | MFCD00866158 |
| InChI Key | BYFMCKSPFYVMOU-UHFFFAOYSA-N |
| Synonym | [(1-Benzyl-3-indazolyl)oxy]acetic Acid, 1-Benzyl-3-[1-(carboxy)methoxy]indazole |
| PubChem CID | 2313 |
| ChEBI | CHEBI:31257 |
| IUPAC Name | 2-(1-benzylindazol-3-yl)oxyacetic acid |
| SMILES | C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=N2)OCC(=O)O |
Chemical Identifiers
| 20187-55-7 | |
| 282.299 | |
| BYFMCKSPFYVMOU-UHFFFAOYSA-N | |
| 2313 | |
| 2-(1-benzylindazol-3-yl)oxyacetic acid |
| C16H14N2O3 | |
| MFCD00866158 | |
| [(1-Benzyl-3-indazolyl)oxy]acetic Acid, 1-Benzyl-3-[1-(carboxy)methoxy]indazole | |
| CHEBI:31257 | |
| C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=N2)OCC(=O)O |
Specifications
| 20187-55-7 | |
| White | |
| MFCD00866158 | |
| [(1-Benzyl-3-indazolyl)oxy]acetic Acid, 1-Benzyl-3-[1-(carboxy)methoxy]indazole | |
| C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=N2)OCC(=O)O | |
| 282.299 | |
| CHEBI:31257 | |
| ≥97.0% (GC,T) | |
| Bendazac |
| 162°C | |
| C16H14N2O3 | |
| 1 g | |
| BYFMCKSPFYVMOU-UHFFFAOYSA-N | |
| 2-(1-benzylindazol-3-yl)oxyacetic acid | |
| 2313 | |
| 282.30 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : AF5085000
TSCA : No