missing translation for 'onlineSavingsMsg'
Learn More

Bendazac 97.0+%, TCI America™

Catalog No. B42231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42231G 1 g
B42235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42231G Supplier TCI America Supplier No. B42231G

Chemical Identifiers

CAS 20187-55-7
Molecular Formula C16H14N2O3
Molecular Weight (g/mol) 282.299
MDL Number MFCD00866158
InChI Key BYFMCKSPFYVMOU-UHFFFAOYSA-N
Synonym [(1-Benzyl-3-indazolyl)oxy]acetic Acid, 1-Benzyl-3-[1-(carboxy)methoxy]indazole
PubChem CID 2313
ChEBI CHEBI:31257
IUPAC Name 2-(1-benzylindazol-3-yl)oxyacetic acid
SMILES C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=N2)OCC(=O)O

Specifications

Melting Point 162°C
Color White
Quantity 1 g
Formula Weight 282.30
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bendazac
RTECSNumber AF5085000
TSCA No