Learn More
Fisherbrand™ Bench Fume Hood Adjustable Height Ceiling Enclosure
Bench Fume Hood Adjustable Height Ceiling Enclosure is designed to be used with Fisherbrand bench Fume Hoods.
 View more versions of this product
Catalog No. 06000623
Only null left
Description
Description
  • These three sided ceiling enclosures are designed to fill the space between the top of a hood and the ceiling up to 24 in. in height to provide a clean finished appearance.
  • The enclosure can be cut to fit and includes a slip to fit ceiling finish trim angle.
Specifications
Chemical Identifiers
Fume Hood
T8 Fluorescent
Fisherbrand Lab Furnishings
35 in.
Fisherbrand Bench Fume Hoods
61 cm
120 V
Powder
Painted Steel
White
88.9 cm
24 in.
Slip to Fit Ceiling Finish Trim Angle
Specifications
Fume Hood
T8 Fluorescent
Fisherbrand Lab Furnishings
35 in.
35-7/8 x 60 x 24 in.
24 in.
Slip to Fit Ceiling Finish Trim Angle
60 in.
Powder
Painted Steel
White
88.9 cm
Fisherbrand Bench Fume Hoods
61 cm
120 V
152.4 cm
Product Suggestions
Product Suggestions