missing translation for 'onlineSavingsMsg'
Learn More

Benazepril hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015705
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015705 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015705 Supplier Merck Emd Millipore Supplier No. PHR1912250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 86541-74-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C24H28N2O5 · HCl
MDL Number MFCD00895734
Synonym Benazepril hydrochloride; (3 S)-3-[[(1 S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1 H-1-benzazepine-1-acetic acid hydrochloride
Type Certified Reference Material
Quantity 250 mg
Formula Weight 460.95
Grade Certified Reference Material