missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Switch-Grid™ Test Tube Rack Grids

Replacement grids

Specifications

Autoclavable Autoclavable
Material Plastic
Shape Rectangular, Rounded Corners
Stackable Yes
Sterility Non-sterile
View More Specs

Products 1
Catalog Number Mfr. No. Holds Array Positions Color Price Quantity & Availability  
Catalog Number Mfr. No. Holds Array Positions Color Price Quantity & Availability  
11123992
View Documents Promotion Details
Bel Art™
F187451300
Tubes (10 to 13 mm) 7 x 6 84 Orange N/A
Only null left
 
Description

Description

Replacement grids

  • Chemical resistant polyoxymethylene plastic
  • Can be cleaned with mild detergent, ethanol or isopropyl alcohol
  • Autoclavable at121°C (250°F)
Specifications

Specifications

Autoclavable
Rectangular, Rounded Corners
Non-sterile
Grid Set
5.6 mm
Plastic
Yes
133 mm
None
102 mm
Videos
SDS
Documents

Documents