Learn More
Bel-Art™ Urbanti™ High-Speed Funnels
Optimized for nonvacuum, high-speed filtration, these glass-clear funnels have internal helicoid ribs to increase the filtration speed
Supplier:  Bel-Art™ F146480000
 View more versions of this product
Catalog No. 10371141
Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left
Description
Description
  • Vertical outer ribs prevent airlock
  • Made from autoclavable polymethylpentene
  • Unaffected by corrosive liquids
  • Heat resistant to 177°C (350°F)
Specifications
Chemical Identifiers
PMP
Autoclavable
Internal Helicoid Ribs
Filling Funnel (Liquid)
Specifications
14 cm
1.6 cm
25.4 cm
24 to 27 cm
13.2 cm
586 mL
PMP
Autoclavable
Internal Helicoid Ribs
Filling Funnel (Liquid)
Product Suggestions
Product Suggestions