missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Bench-Top Biohazard Bag Holder

Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.

Supplier:  Bel-Art™ H13193 1000

 View more versions of this product

Catalog No. 018152


Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left

Includes: Rectangular stainless steel wire frame only

Description

Description

Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.

  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812

Order Info

For the bag holder, cover and biohazard bags, use Cat. No. 11-394-310.

Compliance

When used with lid, provides compliance with 29 CFR 1910.1030(d)(2)(xiii) (A), which requires a cover on any container used to store, transport or ship biohazardous wastes.

Specifications

Chemical Identifiers

Autoclavable
Single-use
5 in.
12.7 cm

Specifications

Epoxy coated
Orange
Perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
Rectangular Stainless-Steel Wire Frame Only
Benchtop
5 in.
21.59 cm
8.63 cm
Autoclavable
Single-use
01-828A, 01-828-031, 01-830A, 01-826-1, 01-827A, 01-829A, 01-815 11 H x 8.5 in. W Biohazard Bags biohazard bag, Clavies™ autoclavable bags (01-815-6)
Steel Wire
3.4 x 5 x 8.5 in. (8.63 x 12.7 x 21.59 cm)
8.5 in.
Biohazard Bag Holder
12.7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions