missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Bench-Top Biohazard Bag Holder
Description
Includes
Plastic Cover Only
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
- Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
- Available with epoxy coat in orange color
- Comes with snug fitting polypropylene plastic cover to hold bag securely in place
- Reusable and allows cleaning by hand or in automated dish washers
- Autoclavable
- Also used with Clavies Autoclavable Bags H13182-0812
Order Info
For complete kit with bag holder, cover and biohazard bags, see Cat. No. 11-394-310.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Orange |
| Disposable | Single-use |
| For Use With (Application) | Store, transport, or ship biohazardous wastes |
| For Use With (Equipment) | Biohazard bag holder |
| Includes | Snug-Fitting Plastic Cover Only |
| Material | Polypropylene |
| Width (English) | 5 in. |
| Product Type | Bag Holder Cover |
| Width (Metric) | 12.7 cm |