Learn More
Bel-Art™ Additional Mesh Inserts
Catalog No. 14306B
Change view
Click to view available options
Screen Size:
Set of 8 (25 Mesh, 35 Mesh, 45 Mesh, 60 Mesh, 80 Mesh, 120 Mesh, 170 Mesh, 230 Mesh)
Catalog No. Screen Size
14306B Set of 8 (25 Mesh, 35 Mesh, 45 Mesh, 60 Mesh, 80 Mesh, 120 Mesh, 170 Mesh, 230 Mesh)
1 options
Catalog No. 14306B Supplier Bel-Art™ Supplier No. F378460000
Get Up to $500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Mini-Sieve™ micro sieve set.
  • Made from phosphor-bronze
  • Removable
  • Includes pack of 8 phosphor-bronze screen inserts, 1 each of 25, 35, 45, 60, 80, 120, 170 and 230 standard mesh
Specifications
Description Removable
Product Type Sieve Insert
Diameter (English) Outer 2.2 in.
For Use With (Equipment) Mini-Sieve micro sieve set
Includes Pack of 8 phosphor-bronze screen inserts, 1 each of 25, 35, 45, 60, 80, 120, 170 and 230 standard mesh
Diameter (Metric) Outer 56 mm
Frame Material Plastic
Screen Size Set of 8 (25 Mesh, 35 Mesh, 45 Mesh, 60 Mesh, 80 Mesh, 120 Mesh, 170 Mesh, 230 Mesh)
Mesh Material Phosphor Bronze
Sieve Number Set of 8 (25, 35, 45, 60, 80, 120, 170, and 230)