Learn More
Bel-Art™ SP Scienceware™ Lab Drawer Compartment Tray
Catalog No. s478862
Change view
Click to view available options
Gadget organizer tray
Catalog No.
S478862 Gadget organizer tray
1 options
Catalog No. S478862 Supplier Bel-Art™ Supplier No. H18652 EMD
Only null left
Invaluable for storing small items.
  • 12 compartments
  • Each compartment is 3.9L x 3.9W x 2.9 in.D (98 x 98 x 73mm)
  • Tray dimensions: 36 x 44.5cm (14 x 17.5 in.); fit standard lab bench drawers
  • White polystyrene plastic
Specifications
Product Type Gadget Organizer Tray
Color White
Material Polystyrene