Learn More
Bel-Art™ Glove Box Economy Sleeve Gloves
One piece, bonded neoprene protection against liquid or gas penetration
Supplier: Bel-Art™ H500250544
Description
- Single piece glove with sleeve
- For glove boxes with 8 in. glove ports
- Requires clamping rings (19-821-983) to hold gloves in place
- Sold in pairs
Specifications
| Black | |
| No | |
| Glove Box Gloves |
| Specific | |
| Bonded Neoprene | |
| 10 |