Learn More
Bel-Art™ Glove Box Economy Sleeve Gloves
Catalog No. p-3273579
Change view
Click to view available options
Size:
10
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19821985 9
19821984 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19821985 Supplier Bel-Art™ Supplier No. H500250546
One piece, bonded neoprene protection against liquid or gas penetration
  • Single piece glove with sleeve
  • For glove boxes with 8 in. glove ports
  • Requires clamping rings (19-821-983) to hold gloves in place
  • Sold in pairs
Specifications
Color Black
Hand Specific
Latex Free No
Material Bonded Neoprene
Product Type Glove Box Gloves
Size 9