missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Wash Bottle, Deionized Water
Right-to-Know Safety-Labeled wash bottles.
Supplier: Bel-Art™ F117160003
Description
- Wide mouth of 53 mm (2 in.) for easy refilling
- Pre-printed with Chemical name and formula, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Translucent LDPE bottles
- Color-coded polypropylene closure
Specifications
| 500 mL | |
| Translucent, Blue | |
| LDPE | |
| Non-sterile | |
| Yes | |
| No | |
| Cylindrical | |
| Deionized Water | |
| Not Autoclavable |
| 16 oz. | |
| Pack of 4 each of “DI Water” wash botttles | |
| 4/Pk. | |
| Blue | |
| Reusable | |
| Wide, Threaded | |
| Wash Bottle | |
| Polypropylene |