missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Cryo-Safe™ Microcentrifuge Tube and Cryogenic Vial Coolers

Benchtop-sized coolers for microcentrifuge tubes or cryogenic vials.

Supplier:  Bel-Art™ F188461000

 View more versions of this product

Catalog No. 03-410-499


Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left

Description

Description

Benchtop-sized coolers for microcentrifuge tubes or cryogenic vials.

  • Holds Twelve 0.5 or 1.5mL tube in individual wells below 0 deg.C for 2.5 Hours
  • Body of cooler filled with a nontoxic gel that does not contact the contents of the cooler
  • Excellent for safe transport of temperature sensitive tubes
  • Polycarbonate body and lid is easy to clean
  • Stackable design with rubber feet to prevent sliding
  • Mini Coolers feature molded-in side handles, Junior and Maxi Coolers have fold down wire carrying handle that helps retain lid
  • Store -15°C coolers at -15°C for 24 hours prior to use
  • Store 0°C coolers at -20°C for a few hours prior to use
  • Store -20°C coolers at -20°C for 24 hours prior to use
Specifications

Specifications

19.7 cm
19.7 cm
14 cm
7.75 x 5.5 x 7.5 in.
1.6 kg
Mini Cooler
15 mL
Autoclavable
0°C
7.5 in.
7.75 in.
5.5 in.
3.5 lb.
Polycarbonate
Clear, Red
12 Microcentrifuge Tubes (15 mL)
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions