missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Conway Diffusion Cells

Catalog No. 0876416
Change view
Click to view available options
Diameter (Metric):
83 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
0876416 83 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0876416 Supplier Bel-Art™ Supplier No. F409410000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Obrink method of determining carbon dioxide, ammonia or alcohol in blood or serum by diffusion method. Bel Art™ Conway Diffusion Cells are steam autoclavable at 121°C (250°F).

  • Smooth, white molded polypropylene with natural, clear polypropylene cover
  • The cover has a lifting knob and fits the annular moat of the cell to seal off the chamber
  • Steam autoclavable at 121°C (250°F)

Specifications

Closure Included Closure Included
Autoclavable Autoclavable
Bottom Shape Flat
Capacity (Metric) 32 mL
Color White
Culture Area 10 x 60 mm
Disposable Reusable
Material Polypropylene
Shape Round
Sterility Non-sterile
Type Conway Diffusion Cell
Volume (Metric) Working 10 x 60 mm
Well Count 1
For Use With (Application) To assay Carbon di Oxide, Ammonia or Alcohol levels in Blood or Serum
Includes Cover
Quantity 3/Pk.
Height (Metric) 10 mm
Depth (English) Center 0.15 in.
Depth (English) Chamber 0.03 in.
Depth (English) Moat 0.15 in.
Depth (Metric) Center 0.4 cm
Depth (Metric) Chamber 0.1 cm
Depth (Metric) Moat 0.4 cm
Diameter (Metric) Center 3.5 cm
Diameter (Metric) Chamber 0.6 cm
Diameter (Metric) 83 mm
Show More Show Less