missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Clear Graduated Cylinders

No concave meniscus, makes the raised numeral, single scale, graduations simple to interpret.

Supplier:  Bel-Art™ F286960000

 View more versions of this product

Catalog No. 14-513-321



Explore available promotions

Description

Description

  • Made from clear TPX
  • Shatter-proof
  • Wide octagonal base prevents tipping and rolling
  • Chemical and heat resistant to 177°C (350°F)
  • Steam autoclavable at 121°C (250°F)

Specifications

Specifications

Plastic
Single
Autoclavable
Round
17.7 in.
2.6 in.
10 mL
10mL
Octagonal
Graduated Cylinder, 1000 mL
SDS
Documents

Documents

Promotions

Promotions